missing translation for 'onlineSavingsMsg'
Learn More

DL-Homocystine, TRC

Product Code. 30397588
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30397588 5 g 5g
30406797 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30397588 Supplier Toronto Research Chemicals Supplier No. TRCH5912905G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 870-93-9
Molecular Formula C8 H16 N2 O4 S2
Molecular Weight (g/mol) 268.35
Synonym DL-4,4'-Dithiobis(2-aminobutyric acid)
IUPAC Name 2-amino-4-[(3-amino-3-carboxy-propyl)disulfanyl]butanoic acid
SMILES NC(CCSSCCC(N)C(=O)O)C(=O)O

Specifications

InChI Formula InChI=1S/C8H16N2O4S2/c9-5(7(11)12)1-3-15-16-4-2-6(10)8(13)14/h5-6H,1-4,9-10H2,(H,11,12)(H,13,14)
Quantity 5 g
Formula Weight 268.0551
Shipping Condition Room Temperature
Chemical Name or Material DL-Homocystine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.