missing translation for 'onlineSavingsMsg'
Learn More

DL-Homocysteine thiolactone hydrochloride, MedChemExpress

Product Code. 16430759
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16430759 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16430759 Supplier Medchem Express Supplier No. HY101404
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress DL-Homocysteine thiolactone hydrochloride is a cyclic amino acid derivative that exhibits root-growth inhibitory activity.

DL-Homocysteine thiolactone hydrochloride is a cyclic amino acid derivative that exhibits root-growth inhibitory activity.

Specifications

Chemical Name or Material DL-Homocysteine thiolactone hydrochloride
Physical Form Powder
CAS 6038-19-3
Color Off-White
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C4H8ClNOS
Solubility Information DMSO : 100 mg/mL (650.91 mM; Need ultrasonic)
SMILES O=C1C(N)CCS1.Cl
Molecular Weight (g/mol) 153.63
Formula Weight 153.63
Percent Purity 97.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.