missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress DL-Homocysteine

Product Code. 16439406
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16439406 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16439406 Supplier MedChemExpress Supplier No. HYW040821
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress DL-Homocysteine is a weak neurotoxin, and can affect the production of kynurenic acid in the brain.

Specifications

Content And Storage Powder -20°C 3 years∣In solvent -80°C 6 months, -20°C 1 month
Molecular Weight (g/mol) 135.19 g/mol
Shipping Condition Room temperature
For Use With (Application) Metabolism-protein/nucleotide metabolism
CAS 454-29-5
Product Type Natural Products
Solubility H2O : 7.14 mg/mL (52.81 mM∣ultrasonic and warming and heat to 80°C)
Target Endogenous Metabolite
Quantity 100 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.