missing translation for 'onlineSavingsMsg'
Learn More

DL-Homocysteic acid, 98%, Thermo Scientific™

Product Code. 15408899
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 15408899

Brand: Thermo Scientific Alfa Aesar L09122.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 504-33-6
Melting Point ∽273°C (decomposition)
Odor Odorless
Molecular Formula C4H9NO5S
MDL Number MFCD00007536
Quantity 1 g
Beilstein 1783391
Synonym dl-homocysteic acid, homocysteic acid, dl-homocysteicacid, 2-amino-4-sulfobutyric acid, d,l-homocysteic acid, pubchem12443, acmc-1ap11, 2-amino-4-sulfo-butanoic acid, dl-2-amino-4-sulfobutyric acid
InChI Key VBOQYPQEPHKASR-UHFFFAOYSA-N
SMILES C(CS(=O)(=O)O)C(C(=O)O)N
IUPAC Name 2-amino-4-sulfobutanoic acid
Molecular Weight (g/mol) 183.178
PubChem CID 92117
Formula Weight 183.18
Percent Purity 98%
Chemical Name or Material DL-Homocysteic acid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.