missing translation for 'onlineSavingsMsg'
Learn More

DL-Histidine hydrochloride, monohydrate, 98%

Product Code. 10055770 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass Bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10055770 100 g Glass Bottle 100g
10444121 25 g Glass Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10055770 Supplier Thermo Scientific Acros Supplier No. 411721000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 123333-71-1 | C6H10ClN3O2 | 191.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123333-71-1
Molecular Formula C6H10ClN3O2
Molecular Weight (g/mol) 191.62
MDL Number MFCD00064555
InChI Key QZNNVYOVQUKYSC-JEDNCBNOSA-N
Synonym hydrogen dl-histidine chloride, 2-amino-3-1h-imidazol-5-yl propanoic acid; hydron; chloride, 2-azanyl-3-1h-imidazol-5-yl propanoic acid; hydron; chloride
PubChem CID 53395185
IUPAC Name 2-amino-3-(1H-imidazol-5-yl)propanoic acid;hydron;chloride
SMILES [H+].[Cl-].N[C@@H](CC1=CN=CN1)C(O)=O

Specifications

Melting Point 233.0°C
Arsenic (As) 1ppm max.
Color White
Heavy Metals (as Pb) 10ppm max.
Infrared Spectrum Authentic
Iron (Fe) 10ppm max.
Assay Percent Range 98%
Loss on Drying 1% max.
Other Amino Acids (TLC) none
Packaging Glass Bottle
Sulfate 300ppm max.
Transmittance 96% min. (c=3, H2O, 430nm) 1cm cell
Sulfated Ash 0.1% max.
Quantity 100 g
Formula Weight 209.64
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material DL-Histidine hydrochloride, monohydrate, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.