missing translation for 'onlineSavingsMsg'
Learn More

Dl-Glyceric Acid Hemicalcium Salt Hydrate, TRC

Product Code. 30388113
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. Quantity unitSize
30388113 1 g 1g
1 options
This item is not returnable. View return policy

Product Code. 30388113

Brand: Toronto Research Chemicals TRCD5260581G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dl-Glyceric Acid Hemicalcium Salt Hydrate
CAS 67525-74-0
Molecular Formula 2 C3 H5 O4 . Ca . 2 H2 O
Quantity 1 g
InChI Formula InChI=1S/2C3H6O4.Ca.2H2O/c2*4-1-2(5)3(6)7;;;/h2*2,4-5H,1H2,(H,6,7);;2*1H2/q;;+2;;/p-2
SMILES O.O.[Ca+2].OCC(O)C(=O)[O-].OCC(O)C(=O)[O-]
IUPAC Name calcium;2,3-dihydroxypropanoate;dihydrate
Molecular Weight (g/mol) 286.247
Formula Weight 286.021
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.