missing translation for 'onlineSavingsMsg'
Learn More

Dl-Glyceric Acid Hemicalcium Salt Hydrate, TRC

Product Code. 30369710
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Product Code. Quantity unitSize
30369710 500 mg 500mg
1 options
This item is not returnable. View return policy

Product Code. 30369710

Brand: Toronto Research Chemicals TRCD526058500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dl-Glyceric Acid Hemicalcium Salt Hydrate
CAS 67525-74-0
Molecular Formula 2 C3 H5 O4 . Ca . 2 H2 O
Quantity 500 mg
InChI Formula InChI=1S/2C3H6O4.Ca.2H2O/c2*4-1-2(5)3(6)7;;;/h2*2,4-5H,1H2,(H,6,7);;2*1H2/q;;+2;;/p-2
SMILES O.O.[Ca+2].OCC(O)C(=O)[O-].OCC(O)C(=O)[O-]
IUPAC Name calcium;2,3-dihydroxypropanoate;dihydrate
Molecular Weight (g/mol) 286.247
Formula Weight 286.021
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.