missing translation for 'onlineSavingsMsg'
Learn More

DL-glutamine, TRC

Product Code. 30404334
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30404334 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30404334 Supplier Toronto Research Chemicals Supplier No. TRCG597115100MG
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material DL-glutamine
CAS 6899-04-3
Molecular Formula C5H10N2O3
Quantity 100 mg
Synonym Glutamine (8CI, 9CI, ACI), (±)-Glutamine, 2-Amino-4-carbamoylbutanoic acid, 2-Amino-4-carbamoylbutyric acid, DL-Gln, DL-Glutamine, NSC 97925, γ-Glutamine
InChI Formula InChI=1S/C5H10N2O3/c6-3(5(9)10)1-2-4(7)8/h3H,1-2,6H2,(H2,7,8)(H,9,10)
SMILES NC(CCC(=O)N)C(=O)O
IUPAC Name 2,5-diamino-5-oxopentanoic acid
Molecular Weight (g/mol) 146.14
Formula Weight 146.0691
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.