missing translation for 'onlineSavingsMsg'
Learn More

DL-Glutamine, MedChemExpress

Product Code. 16488736
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16488736 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16488736 Supplier Medchem Express Supplier No. HYB1346
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress DL-Glutamine is used for biochemical research and drug synthesis.

DL-Glutamine is used for biochemical research and drug synthesis.

Specifications

Chemical Name or Material DL-Glutamine
CAS 6899-04-3
Color White to Off-White
Purity Grade Notes Research
Molecular Formula C5H10N2O3
Synonym (±)-Glutamine DL-Gl
Solubility Information H2O : 33.33 mg/mL (228.07 mM; Need ultrasonic) ∣DMSO : < 1 mg/mL (insoluble or slightly soluble)
SMILES NC(CCC(N)=O)C(O)=O
Molecular Weight (g/mol) 146.14
Quantity 100 mg
Formula Weight 146.14
Percent Purity 97.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.