missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals DL-Glutamic acid monohydrate, 98+%

Product Code. p-7023808
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11473308 25 g 25g
11483308 100 g 100g
11493308 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11473308 Supplier Thermo Scientific Chemicals Supplier No. A17719.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 19285-83-7 | C5H11NO5 | 165.145 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19285-83-7
Molecular Formula C5H11NO5
Molecular Weight (g/mol) 165.145
MDL Number MFCD00150703
InChI Key OZDAOHVKBFBBMZ-UHFFFAOYSA-N
Synonym dl-glutamic acid monohydrate, 2-aminopentanedioic acid hydrate, dl-glutamic acid hydrate, h-dl-glu-oh.h2o, glutaminsaeure hydrate, +/--2-aminoglutaric acid, dl-2-aminopentanedioic acid, 2-azanylpentanedioic acid hydrate, 2-aminopentanedioic acid, oxamethane, hydrate, a-aminoglutarate
PubChem CID 16219385
IUPAC Name 2-aminopentanedioic acid;hydrate
SMILES C(CC(=O)O)C(C(=O)O)N.O

Specifications

Melting Point ∼200°C (decomposition)
Beilstein 4155097
Merck Index 14,4469
Density 1.46
Quantity 25 g
Formula Weight 165.15 (147.13 Anhydrous)
Percent Purity 99%
Chemical Name or Material DL-Glutamic acid monohydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.