missing translation for 'onlineSavingsMsg'
Learn More

DL-Fenfluramine Hydrochloride, TRC

Product Code. 30402853
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30402853 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30402853 Supplier Toronto Research Chemicals Supplier No. TRCF245605250MG

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fenfluramine Hydrochloride
CAS 404-82-0
Purity Grade Notes HPLC
Molecular Formula C12 H16 F3 N . Cl H
Quantity 250 mg
Synonym Fenfluramine hydrochloride, Benzeneethanamine, N-ethyl-α-methyl-3-(trifluoromethyl)-, hydrochloride (1:1), Benzeneethanamine, N-ethyl-α-methyl-3-(trifluoromethyl)-, hydrochloride (9CI), Phenethylamine, N-ethyl-α-methyl-m-(trifluoromethyl)-, hydrochloride (7CI,8CI), (±)-Fenfluramine chloride, (±)-Fenfluramine hydrochloride, 1-(m-Trifluoromethylphenyl)-2-(ethylamino)propane hydrochloride, Ganal, HHR-965, N-Ethyl-α-methyl-m-(trifluoromethyl)phenethylamine hydrochloride, Ponderal, Ponderax, Ponderex, Pondimin, Pondmin, Racemic fenfluramine hydrochloride, S 768, ZX 008, dl-Fenfluramine hydrochloride
InChI Formula InChI=1S/C12H16F3N.ClH/c1-3-16-9(2)7-10-5-4-6-11(8-10)12(13,14)15;/h4-6,8-9,16H,3,7H2,1-2H3;1H
SMILES Cl.CCNC(C)Cc1cccc(c1)C(F)(F)F
IUPAC Name N-ethyl-1-[3-(trifluoromethyl)phenyl]propan-2-amine;hydrochloride
Molecular Weight (g/mol) 267.72
Formula Weight 267.1002
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.