missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-Ethyl Lactate, TRC
Specifications
Specifications
| Chemical Name or Material | Ethyl lactate |
| CAS | 97-64-3 |
| Molecular Formula | C5H10O3 |
| Quantity | 1 g |
| Synonym | Propanoic acid, 2-hydroxy-, ethyl ester, Lactic acid, ethyl ester (6CI,8CI), (±)-Ethyl 2-hydroxypropionate, (±)-Ethyl lactate, (±)-Lactic acid ethyl ester, 2-Hydroxypropanoic acid ethyl ester, Actylol, Acytol, DL-Ethyl lactate, Ethyl 2-hydroxypropanoate, Ethyl 2-hydroxypropionate, Ethyl lactate, Ethyl rac-lactate, Ethyl α-hydroxypropionate, NSC 8850, PBR 40, Purasolv EL, Purasolv ELS, Solactol, Vertec ELS, VertecBio EL, dl-Lactic acid ethyl ester |
| InChI Formula | InChI=1S/C5H10O3/c1-3-8-5(7)4(2)6/h4,6H,3H2,1-2H3 |
| SMILES | CCOC(=O)C(C)O |
| IUPAC Name | ethyl 2-hydroxypropanoate |
| Molecular Weight (g/mol) | 118.13 |
| Formula Weight | 118.063 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?