missing translation for 'onlineSavingsMsg'
Learn More

DL-Ethionine, Approx. 99%, MP Biomedicals™

Product Code. 11498030 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11498030 5 g 5g
11488030 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11498030 Supplier MP Biomedicals Supplier No. 0210166205

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 67-21-0
Molecular Formula C6H13NO2S
Molecular Weight (g/mol) 163.24
MDL Number MFCD00063102
InChI Key GGLZPLKKBSSKCX-UHFFFAOYNA-N
Synonym dl-ethionine, s-ethylhomocysteine, aethionin, s-ethyl-dl-homocysteine, 2-amino-4-ethylthio butyric acid, dl-homocysteine, s-ethyl, ethionine, dl, +--ethionine, homocysteine, s-ethyl, 2-amino-4-ethylthio butanoic acid
PubChem CID 6205
ChEBI CHEBI:68662
IUPAC Name 2-amino-4-(ethylsulfanyl)butanoic acid
SMILES CCSCCC(N)C(O)=O

Specifications

Assay Percent Range Approx. 99%
Quantity 5 g
Formula Weight 163.2
Physical Form Solid
Chemical Name or Material DL-Ethionine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.