missing translation for 'onlineSavingsMsg'
Learn More

DL-Ethionine, 98%

Product Code. 10562921 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2.5 g
10 g
50 g
Packaging:
Glass Bottle
Unit Size:
10g
2.5g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10562921 2.5 g Glass Bottle 2.5g
10794851 10 g Glass Bottle 10g
10259263 50 g Glass Bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10562921 Supplier Thermo Scientific Acros Supplier No. 146170025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 67-21-0 | C6H13NO2S | 163.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-21-0
Molecular Formula C6H13NO2S
Molecular Weight (g/mol) 163.24
MDL Number MFCD00063102
InChI Key GGLZPLKKBSSKCX-UHFFFAOYNA-N
Synonym dl-ethionine, s-ethylhomocysteine, aethionin, s-ethyl-dl-homocysteine, 2-amino-4-ethylthio butyric acid, dl-homocysteine, s-ethyl, ethionine, dl, +--ethionine, homocysteine, s-ethyl, 2-amino-4-ethylthio butanoic acid
PubChem CID 6205
ChEBI CHEBI:68662
IUPAC Name 2-amino-4-ethylsulfanylbutanoic acid
SMILES CCSCCC(N)C(O)=O

Specifications

Melting Point 265°C to 269°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Linear Formula C2H5SCH2CH2CH(NH2)CO2H
Merck Index 15, 3792
Quantity 2.5 g
Formula Weight 163.23
Percent Purity ≥97.5%
Physical Form Crystalline Powder or Flakes
Chemical Name or Material DL-Ethionine
Show More Show Less
compliance-icons
Product Identifier
  • DL-Ethionine
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 2
Hazard Statement
  • H351-Suspected of causing cancer.
  • H361f-Suspected of damaging fertility.
  • H341-Suspected of causing genetic defects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.