missing translation for 'onlineSavingsMsg'
Learn More

DL-Ethionine, 98%

Product Code. 11434534
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11434534 1 g 1g
11444534 5 g 5g
11454534 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11434534 Supplier Thermo Scientific Alfa Aesar Supplier No. A13154.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 67-21-0 | C6H13NO2S | 163.24 g/mol

DL-Ethionine is used as a dietary supplement to accelerate cholangiocarcinogenesis in vivo. It is also used to induce oxidative stress in liver to study the levels and activities of anti-oxidative enzymes and compounds such as glutathione.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-21-0
Molecular Formula C6H13NO2S
Molecular Weight (g/mol) 163.24
MDL Number MFCD00063102
InChI Key GGLZPLKKBSSKCX-UHFFFAOYNA-N
Synonym dl-ethionine, s-ethylhomocysteine, aethionin, s-ethyl-dl-homocysteine, 2-amino-4-ethylthio butyric acid, dl-homocysteine, s-ethyl, ethionine, dl, +--ethionine, homocysteine, s-ethyl, 2-amino-4-ethylthio butanoic acid
PubChem CID 6205
ChEBI CHEBI:68662
SMILES CCSCCC(N)C(O)=O

Specifications

Melting Point ∼265°C (decomposition)
Odor Odorless
Quantity 1 g
Beilstein 1722529
Merck Index 14,3738
Solubility Information Soluble in water.
IUPAC Name 2-amino-4-(ethylsulfanyl)butanoic acid
Formula Weight 163.24
Percent Purity 98%
Chemical Name or Material DL-Ethionine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.