missing translation for 'onlineSavingsMsg'
Learn More

DL-erythro-Dihydrosphingosine, Thermo Scientific™

Product Code. 15494549
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15494549 50 mg 50mg
15484549 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15494549 Supplier Thermo Scientific Alfa Aesar Supplier No. J62103.MF

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
DL-erythro-Dihydrosphingosine is an inhibitor of PKC (protein kinase C). This compound also blocks PLA2 (phospholipases A2) and the D-sphingosine precursor.

Solubility
Soluble in water (partly miscible), chloroform, 100% warm ethanol (25 mg/ml), warm DMSO (25 mg/ml), and methanol.

Notes
Protect from heat. Incompatible with heat and oxidizing agents. Store at -20°C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3102-56-5
Molecular Formula C18H39NO2
Molecular Weight (g/mol) 301.52
MDL Number MFCD00051176,MFCD00079141,MFCD00079141,MFCD00792847
InChI Key OTKJDMGTUTTYMP-UHFFFAOYNA-N
Synonym dl-erythro-dihydrosphingosine, dl-sphinganine, 2r,3s-2-aminooctadecane-1,3-diol, dl-erythro-1,3-dihydroxy-2-aminooctadecane, lopac-d-6908, l-erythro-dihydrosphingosine, dl-erythro-dihydrosphingosine, synthetic
PubChem CID 6603822
IUPAC Name (2R,3S)-2-aminooctadecane-1,3-diol
SMILES CCCCCCCCCCCCCCCC(O)C(N)CO

Specifications

Color White
Quantity 50 mg
Beilstein 1724234
Solubility Information Soluble in water (partly miscible),chloroform,100% warm ethanol (25mg/ml),warm DMSO (25mg/ml),and methanol.
Formula Weight 301.51
Physical Form Powder
Chemical Name or Material DL-erythro-Dihydrosphingosine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.