missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| CAS | 27565-41-9 |
| Molecular Formula | C4H10O2S2 |
| MDL Number | MFCD00004877,MFCD00064305 |
| Quantity | 100 g |
| Synonym | 1, 4-Dithiothreitol, DTT |
| InChI Key | VHJLVAABSRFDPM-UHFFFAOYNA-N |
| SMILES | OC(CS)C(O)CS |
| IUPAC Name | 1,4-disulfanylbutane-2,3-diol |
| Molecular Weight (g/mol) | 154.24 |
| Formula Weight | 154.2 |
| Show More |
Safety and Handling
Product Identifier
- DL-Dithiothreitol
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contain: DL-Dithiothreitol
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?