missing translation for 'onlineSavingsMsg'
Learn More

DL-Dithiothreitol, ≥99.0%, MP Biomedicals™

Product Code. 11477690 Shop All MP Biomedicals Products
Click to view available options
Quantity:
5 g
10 g
25 g
50 g
Unit Size:
10g
25g
50g
5g
This item is not returnable. View return policy

Product Code. 11477690

Brand: MP Biomedicals 0210059725

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 27565-41-9
Molecular Formula C4H10O2S2
Molecular Weight (g/mol) 154.24
MDL Number MFCD00004877,MFCD00064305
InChI Key VHJLVAABSRFDPM-UHFFFAOYNA-N
Synonym DTT, 1, 4-Dithio-DL-threitol, Cleland Reagent
IUPAC Name 1,4-disulfanylbutane-2,3-diol
SMILES OC(CS)C(O)CS

Specifications

Color White
Assay Percent Range ≥99.0%
Quantity 25 g
Formula Weight 154.25
Physical Form Powder
Chemical Name or Material DL-Dithiothreitol
compliance-icons
Product Identifier
  • DL-Dithiothreitol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : DL-Dithiothreitol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.