missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-Buthionine-(S,R)-sulfoximine, MedChemExpress
Description
DL-Buthionine-(S,R)-sulfoximine is a potent inhibitor of glutamylcysteine synthetase biosynthesis.
Specifications
Specifications
| Chemical Name or Material | DL-Buthionine-(S,R)-sulfoximine |
| Physical Form | Solid |
| CAS | 5072-26-4 |
| Color | Off-White |
| Quantity | 50 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C8H18N2O3S |
| For Use With (Application) | Cancer-programmed cell death |
| Synonym | Buthionine sulfoximine BSO |
| Solubility Information | H2O : 25 mg/mL (112.46 mM; ultrasonic and warming and heat to 60°C) |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?