missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals DL-Aspartic acid, 98+%

Product Code. 11410234
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11410234 100 g 100g
11420234 500 g 500g
11430234 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11410234 Supplier Thermo Scientific Chemicals Supplier No. A13646.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 617-45-8 | C4H7NO4 | 133.10 g/mol

DL-Aspartic acid is used in studies on factors and conditions that enhance semen and sperm quality. Its conjugate base D-aspartate has potential use as a therapeutic agent in the treatment of schizophrenia-related symptoms.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-Aspartic acid is used in studies on factors and conditions that enhance semen and sperm quality. Its conjugate base D-aspartate has potential use as a therapeutic agent in the treatment of schizophrenia-related symptoms.

Solubility
Soluble in water, and hydrochloric acid. Insoluble in ether, and alcohol.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 617-45-8
Molecular Formula C4H7NO4
Molecular Weight (g/mol) 133.10
MDL Number MFCD00063083
InChI Key CKLJMWTZIZZHCS-UHFFFAOYNA-N
Synonym dl-aspartic acid, 2-aminosuccinic acid, dl-aminosuccinic acid, dl-asparagic acid, aspartic acid, dl, dl-asp-oh, +--aspartic acid, r,s-aspartic acid, h-dl-asp-oh, acid d,l-aspart
PubChem CID 424
ChEBI CHEBI:22660
IUPAC Name 2-aminobutanedioic acid
SMILES NC(CC(O)=O)C(O)=O

Specifications

Melting Point >300°C (decomposition)
Beilstein 774618
Solubility Information Soluble in water,and hydrochloric acid. Insoluble in ether,and alcohol.
Quantity 100 g
Formula Weight 133.1
Percent Purity ≥98%
Chemical Name or Material DL-Aspartic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.