missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals DL-Asparagine monohydrate, 98%

Catalog Number 10046303
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
This item is not returnable. View return policy
3130-87-8
C4H8N2O3
132.12
MFCD00151039,MFCD00008036,MFCD00064400
DCXYFEDJOCDNAF-UHFFFAOYNA-N
dl-asparagine, asparagin, asparagine, dl, 2-amino-3-carbamoylpropanoic acid, wln: zv1yzvq, h-dl-asn-oh, asparagine #, l-+-asparagine, l-.beta.-asparagine, asparagina
522362
2,4-diamino-4-oxobutanoic acid;hydrate
NC(CC(N)=O)C(O)=O
This item is not returnable. View return policy
3130-87-8
C4H8N2O3
132.12
MFCD00151039,MFCD00008036,MFCD00064400
DCXYFEDJOCDNAF-UHFFFAOYNA-N
dl-asparagine, asparagin, asparagine, dl, 2-amino-3-carbamoylpropanoic acid, wln: zv1yzvq, h-dl-asn-oh, asparagine #, l-+-asparagine, l-.beta.-asparagine, asparagina
522362
2,4-diamino-4-oxobutanoic acid;hydrate
NC(CC(N)=O)C(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Assay Percent Range 98%
Infrared Spectrum Authentic
Linear Formula H2NCOCH2CH(NH2)CO2H·H2O
Beilstein 04, 484
Merck Index 14, 837
Packaging Plastic Bottle
Formula Weight 150.14
Transmittance 90% min. (c=10, 2N HCl, 430nm) 1cm cell
Percent Purity ≥97.5%
Water 10.5 to 13.5% (K.F.)
Quantity 2.5 kg
Physical Form Fine Crystalline Powder
Chemical Name or Material DL-Asparagine monohydrate, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'