missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals DL-Asparagine monohydrate, 98%

Product Code. 10052170 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Unit Size:
100g
2.5kg
25g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10052170 500 g 500g
10741441 25 g 25g
10488910 100 g 100g
10046303 2.5 kg 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10052170 Supplier Thermo Scientific Chemicals Supplier No. 167125000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3130-87-8 | C4H8N2O3 | 132.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3130-87-8
Molecular Formula C4H8N2O3
Molecular Weight (g/mol) 132.12
MDL Number MFCD00151039,MFCD00008036,MFCD00064400
InChI Key DCXYFEDJOCDNAF-UHFFFAOYNA-N
Synonym dl-asparagine, asparagin, asparagine, dl, 2-amino-3-carbamoylpropanoic acid, wln: zv1yzvq, h-dl-asn-oh, asparagine #, l-+-asparagine, l-.beta.-asparagine, asparagina
PubChem CID 522362
IUPAC Name 2,4-diamino-4-oxobutanoic acid;hydrate
SMILES NC(CC(N)=O)C(O)=O

Specifications

Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic Bottle
Linear Formula H2NCOCH2CH(NH2)CO2H·H2O
Beilstein 04, 484
Merck Index 14, 837
Transmittance 90% min. (c=10, 2N HCl, 430nm) 1cm cell
Water 10.5 to 13.5% (K.F.)
Quantity 500 g
Formula Weight 150.14
Percent Purity ≥97.5%
Physical Form Fine Crystalline Powder
Chemical Name or Material DL-Asparagine monohydrate, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.