missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ DL-Arginine monohydrochloride monohydrate, 98+%, Thermo Scientific™

Product Code. 11419652
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11419652 5 g 5g
11429652 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11419652 Supplier Alfa Aesar™ Supplier No. A10758.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 332360-01-7
Molecular Formula C6H17ClN4O3
Molecular Weight (g/mol) 228.677
MDL Number MFCD00151031
InChI Key NUFCANQNMWVENN-UHFFFAOYSA-N
Synonym dl-arginine hydrochloride monohydrate, 2-amino-5-guanidinopentanoic acid hydrochloride hydrate, 2-amino-5-diaminomethylideneamino pentanoic acid hydrate hydrochloride, h-dl-arg-oh.hcl, arginine hydrate hydrochloride, pubchem12368, acmc-1ajcb, arginine, chloride, hydrate, dl-arginine monohydrochloride 1-hydrate, dl-arginine monohydrochloride monohydrate
PubChem CID 2734011
IUPAC Name 2-amino-5-(diaminomethylideneamino)pentanoic acid;hydrate;hydrochloride
SMILES C(CC(C(=O)O)N)CN=C(N)N.O.Cl

Specifications

Beilstein 4158960
Quantity 5 g
Formula Weight 228.69 (210.67 Anhydrous)
Percent Purity ≥98%
Chemical Name or Material DL-Arginine monohydrochloride monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.