missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals DL-Arginine hydrochloride

Product Code. 15494029
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15494029 25 g 25g
15484029 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15494029 Supplier Thermo Scientific Chemicals Supplier No. J61420.14
 Restricted Item
Estimated Shipment: 16-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Substrate for nitric oxide synthase; induces insulin release by a nitric oxide-dependent mechanism | CAS: 32042-43-6 | C6H15ClN4O2 | 210.662 g/mol

Substrate for nitric oxide synthase; induces insulin release by a nitric oxide-dependent mechanism

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32042-43-6
Molecular Formula C6H15ClN4O2
Molecular Weight (g/mol) 210.662
MDL Number MFCD00064549
InChI Key KWTQSFXGGICVPE-UHFFFAOYSA-N
Synonym dl-arginine hydrochloride, 2-amino-5-guanidinopentanoic acid hydrochloride, argininhydrochlorid, dl-arginine hcl, h-dl-arg-oh.xhcl, dl-+/--arginine hydrochloride, 1-hydrochloride arginine, arginine monochloride, h-dl-arg-oh.hcl, wln: muyzm3yzvq &gh
PubChem CID 85880
IUPAC Name 2-amino-5-(diaminomethylideneamino)pentanoic acid;hydrochloride
SMILES C(CC(C(=O)O)N)CN=C(N)N.Cl

Specifications

Color White
Quantity 25 g
Formula Weight 210.67
Physical Form Powder
Chemical Name or Material DL-Arginine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.