missing translation for 'onlineSavingsMsg'
Learn More

DL-AP5 Sodium Salt, TRC

Product Code. 30381701
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30381701 10 mg
30370722 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30381701 Supplier Toronto Research Chemicals Supplier No. TRCD52536510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1303993-72-7
Molecular Formula C5H11NNaO5P
Molecular Weight (g/mol) 219.11
Synonym 5-Phosphono-norvaline Sodium Salt
IUPAC Name sodium (S)-2-amino-5-phosphonopentanoate
SMILES N[C@@H](CCCP(O)(O)=O)C(O[Na])=O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C5H12NO5P.Na/c6-4(5(7)8)2-1-3-12(9,10)11;/h4H,1-3,6H2,(H,7,8)(H2,9,10,11);/q;+1/p-1/t4-;/m0./s1
Formula Weight 219.03
Chemical Name or Material DL-AP5 Sodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.