missing translation for 'onlineSavingsMsg'
Learn More

DL-alpha-Tocopherylacetate (Vitamin E acetate) 10 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30325781
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30325781 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30325781 Supplier Dr. Ehrenstorfer Supplier No. DREA17924320AL10
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Vitamin E acetate (DL-alpha-Tocopherol acetate)
CAS 52225-20-4
Analyte or Component Names Vitamin E acetate (DL-alpha-Tocopherol acetate)
Shipping Condition Room Temperature
Molecular Formula C31 H52 O3
Concentration 10 μg/mL
Certifications/Compliance ISO 17034
Synonym 2H-1-Benzopyran-6-ol, 3,4-dihydro-2,5,7,8-tetramethyl-2-(4,8,12-trimethyltridecyl)-, acetate, [2R*(4R*,8R*)]-(±)-, 2H-1-Benzopyran-6-ol, 3,4-dihydro-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-, acetate, (2R)-rel- (9CI), (±)-α-Tocopherol acetate, Alfacol, DL-α-Tocopherol acetate, DL-α-Tocopheryl acetate, Detulin, Diluvac Forte, E-Vimin, Ephynal, Eusovit, Evigen, Fertilvit, Gevex, OptoVit E, Rovimix E, Rovimix E 50, Rovimix E 50AS, Rovimix E 50Ads, Rovimix E 50SD, SynAC, Syntopherol acetate, Toco500, Tocopherol acetate, Vitagutt, all-rac-α-Tocopheryl acetate, dl-Vitamin E acetate, dl-α-Tocopherol acetate, dl-α-Tocopheryl acetate, α-Tocopherol acetate, α-Tocopheryl acetate
InChI Formula InChI=1S/C31H52O3/c1-21(2)13-10-14-22(3)15-11-16-23(4)17-12-19-31(9)20-18-28-26(7)29(33-27(8)32)24(5)25(6)30(28)34-31/h21-23H,10-20H2,1-9H3/t22-,23-,31-/m1/s1
SMILES CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCc2c(C)c(OC(=O)C)c(C)c(C)c2O1
IUPAC Name [(2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydrochromen-6-yl] acetate
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 472.74
Quantity 1 mL
Formula Weight 472.3916 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.