missing translation for 'onlineSavingsMsg'
Learn More

DL-alpha-Tocopherol, 97+%

Product Code. 11408906
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11408906 1000 g 1000g
11418906 50 g 50g
11428906 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11408906 Supplier Thermo Scientific Alfa Aesar Supplier No. A17039.0B
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10191-41-0 | C29H50O2 | 430.72 g/mol

DL-α-Tocopherol is used as an antioxidant that protects cell membrane lipids from oxidative damage.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-α-Tocopherol is used as an antioxidant that protects cell membrane lipids from oxidative damage.

Solubility
Miscible with chloroform, vegetable oils,  ether, acetone and alcohol. Immiscible with water.

Notes
Store in a cool place. Incompatible withstrong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10191-41-0
Molecular Formula C29H50O2
Molecular Weight (g/mol) 430.72
MDL Number MFCD00072051
InChI Key GVJHHUAWPYXKBD-IEOSBIPESA-N
Synonym vitamin e, alpha-tocopherol, d-alpha-tocopherol, 5,7,8-trimethyltocol, +-alpha-tocopherol, r,r,r-alpha-tocopherol, phytogermine, eprolin, 2r,4'r,8'r-alpha-tocopherol, dl-a-tocopherol
PubChem CID 14985
ChEBI CHEBI:18145
IUPAC Name (2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydrochromen-6-ol
SMILES CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCC2=C(C)C(O)=C(C)C(C)=C2O1

Specifications

Melting Point 2°C to 4°C
Density 0.95
Boiling Point 200°C to 220°C (0.1 mmHg)
Flash Point 240°C (464°F)
Assay Percent Range >97%
Refractive Index 1.505
Quantity 1000 g
Beilstein 94012
Sensitivity Air and light sensitive
Merck Index 14,10021
Solubility Information Miscible with chloroform,vegetable oils, ether,acetone and alcohol. Immiscible with water.
Formula Weight 430.72
Percent Purity ≥97%
Chemical Name or Material DL-alpha-Tocopherol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.