missing translation for 'onlineSavingsMsg'
Learn More

DL-alpha-Methylbenzylamine, TRC

Product Code. 30136298
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136298 25 g 25g
30135006 5 g 5g
30005993 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136298 Supplier Toronto Research Chemicals Supplier No. TRCM28901025G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 618-36-0
Molecular Formula C8 H11 N
Molecular Weight (g/mol) 121.18
Synonym Benzenemethanamine, α-methyl-, 1-Phenylethanamine, α-Phenylethylamine
IUPAC Name 1-phenylethanamine
SMILES CC(N)c1ccccc1

Specifications

Quantity 25 g
InChI Formula InChI=1S/C8H11N/c1-7(9)8-5-3-2-4-6-8/h2-7H,9H2,1H3
Formula Weight 121.0891
Percent Purity >95%
Chemical Name or Material DL-alpha-Methylbenzylamine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.