missing translation for 'onlineSavingsMsg'
Learn More

DL-alpha-(Methylaminomethyl)benzyl alcohol, 99%, Thermo Scientific™

Product Code. 11367985
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
This item is not returnable. View return policy

Product Code. 11367985

Brand: Thermo Scientific Alfa Aesar L08111.04

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6589-55-5
Molecular Formula C9H13NO
Molecular Weight (g/mol) 151.209
MDL Number MFCD00004506
InChI Key ZCTYHONEGJTYQV-UHFFFAOYSA-N
Synonym 2-methylamino-1-phenylethanol, halostachine, n-methylphenylethanolamine, +--halostachine, 2-methylamino-1-phenylethan-1-ol, +--alpha-methylamino methyl benzenemethanol, alpha-methylamino methyl benzyl alcohol, ethanol, 2-methylamino-1-phenyl, benzyl alcohol, alpha-methylamino methyl-, dl, alpha-methylaminomethyl benzyl alcohol
PubChem CID 913
ChEBI CHEBI:16913
IUPAC Name 2-(methylamino)-1-phenylethanol
SMILES CNCC(C1=CC=CC=C1)O

Specifications

Melting Point 73°C to 76°C
Quantity 2 g
Beilstein 1072841
Merck Index 14,4601
Formula Weight 151.21
Percent Purity 99%
Chemical Name or Material DL-alpha-(Methylaminomethyl)benzyl alcohol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.