missing translation for 'onlineSavingsMsg'
Learn More

DL-Alanine, TRC

Product Code. 30179396
Change view
Click to view available options
Quantity:
10 g
100 g
250 g
Unit Size:
100g
10g
250g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30179396 100 g 100g
30179142 10 g 10g
30179139 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30179396 Supplier Toronto Research Chemicals Supplier No. TRCA481400100G
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 302-72-7
Molecular Formula C3 H7 N O2
Molecular Weight (g/mol) 89.09
Synonym Alanine, DL- (8CI), DL-Alanine, (R,S)-Alanine, (±)-2-Aminopropionic acid, (±)-Alanine, DL-Ala, DL-α-Alanine, DL-α-Aminopropionic acid, NSC 7602, dl-2-Aminopropanoic acid, dl-Alanine
IUPAC Name 2-aminopropanoic acid
SMILES CC(N)C(=O)O

Specifications

InChI Formula InChI=1S/C3H7NO2/c1-2(4)3(5)6/h2H,4H2,1H3,(H,5,6)
Quantity 100 g
Formula Weight 89.0477
Shipping Condition Room Temperature
Chemical Name or Material DL-Alanine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.