missing translation for 'onlineSavingsMsg'
Learn More

DL-Alanine methyl ester hydrochloride, 98+%

Product Code. 11464324
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11464324 25 g 25g
11454324 5 g 5g
11474324 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11464324 Supplier Thermo Scientific Alfa Aesar Supplier No. A12599.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13515-97-4 | C4H10ClNO2 | 139.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13515-97-4
Molecular Formula C4H10ClNO2
Molecular Weight (g/mol) 139.58
MDL Number MFCD00035523
InChI Key IYUKFAFDFHZKPI-UHFFFAOYNA-N
Synonym methyl 2-aminopropanoate hydrochloride, dl-alanine methyl ester hydrochloride, h-dl-ala-ome.hcl, h-dl-ala-ome hydrochloride, dl-alanine methyl ester hcl, methyl l-alaninate hcl, methyl dl-alaninate hcl, methyl dl-alaninate hydrochloride, h-dl-ala-ome hcl, methyl dl-2-aminopropanoate hydrochloride
PubChem CID 2756298
IUPAC Name methyl 2-aminopropanoate;hydrochloride
SMILES Cl.COC(=O)C(C)N

Specifications

Melting Point ∼155°C (decomposition)
Beilstein 3619264
Quantity 25 g
Formula Weight 139.58
Sensitivity Very Hygroscopic
Percent Purity ≥98%
Chemical Name or Material DL-Alanine methyl ester hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.