missing translation for 'onlineSavingsMsg'
Learn More

DL-Alanine ethyl ester hydrochloride, 99%

Product Code. 11333146
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11333146 5 g 5g
11343146 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11333146 Supplier Thermo Scientific Alfa Aesar Supplier No. L11020.06
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 617-27-6 | C5H12ClNO2 | 153.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 617-27-6
Molecular Formula C5H12ClNO2
Molecular Weight (g/mol) 153.61
MDL Number MFCD00013018
InChI Key JCXLZWMDXJFOOI-UHFFFAOYNA-N
Synonym ethyl 2-aminopropanoate hydrochloride, dl-alanine ethyl ester hydrochloride, h-dl-ala-oet.hcl, h-dl-ala-oet hydrochloride, d,l-alanine ethyl ester hydrochloride, h-dl-ala-oet hcl, l-alanine ethyl ester hcl, alanine, ethyl ester, hydrochloride 1:1, alanine, ethyl ester, hydrochloride, ethyl alaninate hydrochloride
PubChem CID 102560
IUPAC Name ethyl 2-aminopropanoate;hydrochloride
SMILES Cl.CCOC(=O)C(C)N

Specifications

Melting Point 84°C to 86°C
Beilstein 3654425
Quantity 5 g
Formula Weight 153.61
Sensitivity Hygroscopic
Percent Purity 99%
Chemical Name or Material DL-Alanine ethyl ester hydrochloride
compliance-icons
Product Identifier
  • DL-Alanine ethyl ester hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: DL-Alanine ethyl ester hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.