missing translation for 'onlineSavingsMsg'
Learn More

DL-Alanine ethyl ester hydrochloride, 99%, Thermo Scientific Chemicals

Catalog Number p-52459
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
617-27-6
C5H12ClNO2
153.61
MFCD00013018
JCXLZWMDXJFOOI-UHFFFAOYNA-N
ethyl 2-aminopropanoate hydrochloride, dl-alanine ethyl ester hydrochloride, h-dl-ala-oet.hcl, h-dl-ala-oet hydrochloride, d,l-alanine ethyl ester hydrochloride, h-dl-ala-oet hcl, l-alanine ethyl ester hcl, alanine, ethyl ester, hydrochloride 1:1, alanine, ethyl ester, hydrochloride, ethyl alaninate hydrochloride
102560
ethyl 2-aminopropanoate;hydrochloride
Cl.CCOC(=O)C(C)N
This item is not returnable. View return policy
617-27-6
C5H12ClNO2
153.61
MFCD00013018
JCXLZWMDXJFOOI-UHFFFAOYNA-N
ethyl 2-aminopropanoate hydrochloride, dl-alanine ethyl ester hydrochloride, h-dl-ala-oet.hcl, h-dl-ala-oet hydrochloride, d,l-alanine ethyl ester hydrochloride, h-dl-ala-oet hcl, l-alanine ethyl ester hcl, alanine, ethyl ester, hydrochloride 1:1, alanine, ethyl ester, hydrochloride, ethyl alaninate hydrochloride
102560
ethyl 2-aminopropanoate;hydrochloride
Cl.CCOC(=O)C(C)N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 87.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (Argentometry)
Packaging Glass bottle
Linear Formula CH3CH(NH)2COOC2H5·HCl
Quantity 5 g
Formula Weight 153.61
Percent Purity 99%
Physical Form Granular Powder and Granules
Chemical Name or Material DL-Alanine ethyl ester hydrochloride, 99%
Show More Show Less
compliance-icons
Product Identifier
  • DL-Alanine ethyl ester hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'