missing translation for 'onlineSavingsMsg'
Learn More

DL-6-Methoxy-alpha-methyl-2-naphthalenemethanol, 98%

Product Code. 10081691 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10081691 25 g Glass bottle 25g
10244761 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10081691 Supplier Thermo Scientific Acros Supplier No. 331500250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 77301-42-9 | C13H14O2 | 202.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77301-42-9
Molecular Formula C13H14O2
Molecular Weight (g/mol) 202.25
MDL Number MFCD01632581
InChI Key OUVJWFRUESFCCY-SECBINFHSA-N
Synonym 1-6-methoxy-2-naphthyl ethanol, 1-6-methoxynaphthalen-2-yl ethanol, 1-6'-methoxy-2-naphthyl ethanol, +/--1-6-methoxynaphthyl ethanol, 2-1-hydroxyethyl-6-methoxynaphthalene, 1-6-methoxynaphthalen-2-yl ethan-1-ol, 6-methoxy-alpha-methyl-2-naphthalenemethanol, 1rs-1-6-methoxy-2-naphthyl ethanol naproxen impurity k, 1-6-methoxy-2-naphthyl ethan-1-ol, naproxen impurity k
PubChem CID 575523
IUPAC Name 1-(6-methoxynaphthalen-2-yl)ethanol
SMILES COC1=CC=C2C=C(C=CC2=C1)[C@@H](C)O

Specifications

Melting Point 111.0°C to 114.0°C
Color Beige-Yellow to White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in acetone,dichloromethane,ethylacetate,slightly soluble in benzene
Formula Weight 202.25
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material DL-6-Methoxy-α-methyl-2-naphthalenemethanol, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.