missing translation for 'onlineSavingsMsg'
Learn More

DL-6-Hydroxy Norleucine, TRC

Product Code. 30400007
Change view
Click to view available options
Quantity:
500 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30400007 5 g
30404073 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30400007 Supplier Toronto Research Chemicals Supplier No. TRCH9488555G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 305-77-1
Molecular Formula C6 H13 N O3
Molecular Weight (g/mol) 147.17
IUPAC Name 2-amino-6-hydroxyhexanoic acid
SMILES NC(CCCCO)C(=O)O

Specifications

InChI Formula InChI=1S/C6H13NO3/c7-5(6(9)10)3-1-2-4-8/h5,8H,1-4,7H2,(H,9,10)
Quantity 5 g
Formula Weight 147.0895
Shipping Condition Room Temperature
Chemical Name or Material DL-6-Hydroxy Norleucine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.