missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals DL-5-Hydroxytryptophan, 99%

Product Code. 11414933
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11414933 5 g 5g
11404933 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11414933 Supplier Thermo Scientific Chemicals Supplier No. A12237.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 56-69-9 | C11H12N2O3 | 220.23 g/mol

It is an immediate precursor that is used to assess central serotonergic function. It was used for the treatment of insomnia.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an immediate precursor that is used to assess central serotonergic function. It was used for the treatment of insomnia.

Solubility
Soluble in water (4 mg/ml at 25°C), methanol.

Notes
Store at 4°C. Store in cool, dry place in a tightly sealed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-69-9
Molecular Formula C11H12N2O3
Molecular Weight (g/mol) 220.23
MDL Number MFCD00005651
InChI Key LDCYZAJDBXYCGN-UHFFFAOYSA-N
Synonym 5-hydroxy-dl-tryptophan, 5-hydroxytryptophan, dl-5-hydroxytryptophan, 2-amino-3-5-hydroxy-1h-indol-3-yl propanoic acid, +--5-hydroxytryptophan, dl-hydroxytryptophan, dl-5-htp, 5-hydroxytryptophan dl-form, h-dl-trp 5-oh-oh, dl-pretonine
PubChem CID 144
ChEBI CHEBI:28171
IUPAC Name 2-amino-3-(5-hydroxy-1H-indol-3-yl)propanoic acid
SMILES NC(CC1=CNC2=CC=C(O)C=C12)C(O)=O

Specifications

Melting Point ∼298°C (decomposition)
Beilstein 88199
Merck Index 14,4847
Solubility Information Soluble in water (4mg/ml at 25°C),methanol.
Quantity 5 g
Formula Weight 220.23
Percent Purity 99%
Chemical Name or Material DL-5-Hydroxytryptophan

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.