missing translation for 'onlineSavingsMsg'
Learn More

DL-4-Hydroxymandelic acid monohydrate, 97%

Product Code. p-7044334
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11327645 5 g 5g
11337645 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11327645 Supplier Thermo Scientific Alfa Aesar Supplier No. L05916.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 184901-84-6 | C8H10O5 | 186.163 g/mol

DL-4-Hydroxymandelic acid monohydrate is used in the synthesis of an enzyme activity-based labeling molule.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-4-Hydroxymandelic acid monohydrate is used in the synthesis of an enzyme activity-based labeling molule.

Solubility
Soluble in water.

Notes
Incompatible with air, heat and oxidizing agents. Store away from air. Protect from heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 184901-84-6
Molecular Formula C8H10O5
Molecular Weight (g/mol) 186.163
MDL Number MFCD00149283
InChI Key ATPBHLAWGXOMOR-UHFFFAOYSA-N
Synonym 2-hydroxy-2-4-hydroxyphenyl acetic acid hydrate, 4-hydroxymandelic acid monohydrate, dl-4-hydroxymandelic acid monohydrate, 4-hydroxymandelic acid hydrate, 3-hydroxymandelic acid monohydrate, hydroxy 4-hydroxyphenyl acetic acid-water 1/1, hydroxy-4-hydroxy-phenyl-acetic acid monohydrate, benzeneacetic acid, a,4-dihydroxy-, hydrate 1:1
PubChem CID 12677290
IUPAC Name 2-hydroxy-2-(4-hydroxyphenyl)acetic acid;hydrate
SMILES C1=CC(=CC=C1C(C(=O)O)O)O.O

Specifications

Melting Point 88°C to 90°C
Quantity 5 g
Beilstein 2365374
Solubility Information Soluble in water.
Formula Weight 186.17 (168.15 Anhydrous)
Percent Purity 97%
Chemical Name or Material DL-4-Hydroxymandelic acid monohydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.