missing translation for 'onlineSavingsMsg'
Learn More

DL-4-Fluoro-alpha-methylbenzylamine, 97%, Thermo Scientific™

Product Code. 10147303 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10147303 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10147303 Supplier Thermo Scientific Acros Supplier No. 167380010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 403-40-7
Molecular Formula C8H10FN
Molecular Weight (g/mol) 139.17
MDL Number MFCD00041323
InChI Key QGCLEUGNYRXBMZ-UHFFFAOYSA-N
Synonym 1-4-fluorophenyl ethanamine, 1-4-fluorophenyl ethylamine, 4-fluoro-alpha-methylbenzylamine, 1-4'-fluorophenyl ethanamine, dl-4-fluoro-alpha-methylbenzylamine, 4-fluoro-a-methylbenzylamine, 1-p-fluorophenyl-ethylamine, 1-4-fluorophenyl ethan-1-amine, p-fluoro-a-methylbenzylamine, rs-1-4-fluorophenyl ethylamine
PubChem CID 123063
IUPAC Name 1-(4-fluorophenyl)ethanamine
SMILES CC(C1=CC=C(C=C1)F)N

Specifications

Melting Point -30°C
Color Colorless to Yellow or Orange
Density 1.0590g/mL
Boiling Point 97°C (35.0 mmHg)
Flash Point 68°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula FC6H4CH(CH3)NH2
Refractive Index 1.5005 to 1.5025
Quantity 1 g
Specific Gravity 1.059
Formula Weight 139.17
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material DL-4-Fluoro-α-methylbenzylamine, 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.