missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals DL-4-Chlorophenylalanine, 98%

Product Code. 10163060 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass Bottle
Plastic Bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10163060 5 g Glass Bottle 5g
10408130 25 g Plastic Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10163060 Supplier Thermo Scientific Chemicals Supplier No. 157280050
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7424-00-2 | C9H10ClNO2 | 199.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7424-00-2
Molecular Formula C9H10ClNO2
Molecular Weight (g/mol) 199.63
MDL Number MFCD00002601
InChI Key NIGWMJHCCYYCSF-UHFFFAOYNA-N
Synonym fenclonine, 4-chloro-dl-phenylalanine, p-chlorophenylalanine, 2-amino-3-4-chlorophenyl propanoic acid, 4-chlorophenylalanine, dl-4-chlorophenylalanine, dl-p-chlorophenylalanine, fenchlonine, fenclonin, phenylalanine, 4-chloro
PubChem CID 4652
IUPAC Name 2-amino-3-(4-chlorophenyl)propanoic acid
SMILES NC(CC1=CC=C(Cl)C=C1)C(O)=O

Specifications

Melting Point >240°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Linear Formula ClC6H4CH2CH(NH2)CO2H
Beilstein 14, I, 608
Solubility Information Solubility in water: slightly soluble. Other solubilities: slightly soluble in ethanol
Water 0.5% max. (K.F.)
Quantity 5 g
Formula Weight 199.64
Percent Purity 98%
Physical Form Powder
Chemical Name or Material DL-4-Chlorophenylalanine, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.