missing translation for 'onlineSavingsMsg'
Learn More

DL-3-phenyllactic acid, TRC

Product Code. 30404481
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30404481 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30404481 Supplier Toronto Research Chemicals Supplier No. TRCH9537131G
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material DL-3-phenyllactic acid
CAS 828-01-3
Molecular Formula C9H10O3
Quantity 1 g
Synonym a-Hydroxy-benzenepropanoic Acid(±)-, DL-3-phenyl Lactic Acid (8CI), b-Phenyl- Lactic Acid, (2CI), a-Hydroxybenzenepropanoic Acid, (RS)-3-Phenyllactic Acid, (±)-2-Hydroxy-3-phenylpropanoic Acid, (±)-3-Phenyllactic Acid, (±)-b-Phenyllactic Acid, 2-Hydroxy-3-phenylpropanoic Acid, 2-Hydroxy-3-phenylpropionic Acid, 3-Phenyl-2-hydroxypropanoic Acid, 3-Phenyllactic Acid, Ba 2653, DL-2-Hydroxy-3-phenylpropionic Acid, DL-b-Phenyllactic Acid, NSC 2627, dl-Phenyllactic Acid, a-Hydroxy-b-phenylpropionic Acid, a-Hydroxyphenylpropanoic Acid, b-Phenyllactic Acid
InChI Formula InChI=1S/C9H10O3/c10-8(9(11)12)6-7-4-2-1-3-5-7/h1-5,8,10H,6H2,(H,11,12)
SMILES OC(Cc1ccccc1)C(=O)O
IUPAC Name 2-hydroxy-3-phenylpropanoic acid
Molecular Weight (g/mol) 166.17
Formula Weight 166.063
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.