missing translation for 'onlineSavingsMsg'
Learn More

DL-3-Octanol, 97%

Product Code. 10116683 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 mL
250 mL
Packaging:
Glass bottle
Unit Size:
250mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10116683 250 mL Glass bottle 250mL
10194310 50 mL Glass bottle 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10116683 Supplier Thermo Scientific Acros Supplier No. 203772500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 589-98-0 | C8H18O | 130.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 589-98-0
Molecular Formula C8H18O
Molecular Weight (g/mol) 130.23
MDL Number MFCD00004590
InChI Key NMRPBPVERJPACX-UHFFFAOYNA-N
Synonym 3-octanol, octanol-3, amyl ethyl carbinol, ethyl-n-amylcarbinol, amylethylcarbinol, ethylamylcarbinol, 1-ethylhexanol, d-n-octanol, ethyl amyl carbinol, s-3-octanol
PubChem CID 11527
ChEBI CHEBI:80945
IUPAC Name octan-3-ol
SMILES CCCCCC(O)CC

Specifications

Melting Point -45.0°C
Color Colorless
Density 0.8250g/mL
Boiling Point 174.0°C to 176.0°C
Flash Point 67°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)4CH(OH)C2H5
Refractive Index 1.4220 to 1.4280
Quantity 250 mL
Beilstein 01, I, 208
Specific Gravity 0.825
Solubility Information Solubility in water: insoluble.
Formula Weight 130.23
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material DL-3-Octanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.