missing translation for 'onlineSavingsMsg'
Learn More

DL-3-Methylcyclopentanone, 99%

Product Code. 10423035 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10423035 25 g Glass bottle 25g
10183750 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10423035 Supplier Thermo Scientific Acros Supplier No. 169370250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1757-42-2 | C6H10O | 98.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1757-42-2
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.14
MDL Number MFCD00001416
InChI Key AOKRXIIIYJGNNU-UHFFFAOYSA-N
Synonym 3-methylcyclopentanone, cyclopentanone, 3-methyl, dl-3-methylcyclopentanone, 3-methyl-1-cyclopentanone, cyclopentanone, 3-methyl-, r, 3-methylcyclopentanone, +/-, acmc-20apjx, 3-methyl cyclopentanone, 3-methyl-cyclopentanone, acmc-1ba4k
PubChem CID 15650
IUPAC Name 3-methylcyclopentan-1-one
SMILES CC1CCC(=O)C1

Specifications

Color Yellow
Density 0.9100g/mL
Boiling Point 145°C
Flash Point 36°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CH3C5H7(=O)
Refractive Index 1.4330 to 1.435
Quantity 25 g
Beilstein 07, 11
Specific Gravity 0.91
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol,ether and acetone
Formula Weight 98.14
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material DL-3-Methylcyclopentanone
Show More Show Less
compliance-icons
Product Identifier
  • DL-3-Methylcyclopentanone
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.