missing translation for 'onlineSavingsMsg'
Learn More

DL-3-Indolylglycine, MedChemExpress

Product Code. 16482848
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
500mg
50mg
This item is not returnable. View return policy

Product Code. 16482848

Brand: Medchem Express HY100217

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress DL-3-Indolylglycine is an unnatural amino acid that is very similar to Tryptophan, with the indole moiety directly attached to the α-position.

DL-3-Indolylglycine is an unnatural amino acid that is very similar to Tryptophan, with the indole moiety directly attached to the α-position.

Specifications

Chemical Name or Material DL-3-Indolylglycine
CAS 6747-15-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C10H10N2O2
Solubility Information H2O : 4.63 mg/mL (24.34 mM; ultrasonic and warming and adjust pH to 2 with HCl and heat to 60°C)
SMILES NC(C(O)=O)C1=CNC2=CC=CC=C21
Molecular Weight (g/mol) 190.2
Quantity 10 mg
Formula Weight 190.2
Percent Purity 98.16%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.