missing translation for 'onlineSavingsMsg'
Learn More

DL-3-Hydroxybutyric acid, sodium salt, 99%

Product Code. 10107803 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10107803 100 g Glass bottle 100g
10184790 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10107803 Supplier Thermo Scientific Acros Supplier No. 215011000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 150-83-4 | C4H7NaO3 | 126.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 150-83-4
Molecular Formula C4H7NaO3
Molecular Weight (g/mol) 126.09
MDL Number MFCD00016716
InChI Key NBPUSGBJDWCHKC-UHFFFAOYSA-M
Synonym sodium 3-hydroxybutyrate, sodium 3-hydroxybutanoate, dl-3-hydroxybutyric acid sodium salt, sodium beta-hydroxybutyrate, butanoic acid, 3-hydroxy-, monosodium salt, butanoic acid,3-hydroxy-, sodium salt 1:1, dl-3-hydroxybutyric acid, sodium salt, 3-hydroxybutyric acid sodium salt, sodium 3-oxidanylbutanoate, dl-beta-hydroxybutyric acid sodium salt
PubChem CID 23676771
IUPAC Name sodium;3-hydroxybutanoate
SMILES CC(CC(=O)[O-])O.[Na+]

Specifications

Melting Point 161.0°C to 167.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CH3CH(OH)CH2CO2Na
Quantity 100 g
Beilstein 03, 308
Solubility Information Solubility in water: freely soluble
Formula Weight 126.09
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material DL-3-Hydroxybutyric acid, sodium salt
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.