missing translation for 'onlineSavingsMsg'
Learn More

DL-3-Aminoisobutyric Acid, TRC

Product Code. 30323181
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
This item is not returnable. View return policy

Product Code. 30323181

Brand: Toronto Research Chemicals TRCA603043250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material DL-3-Aminoisobutyric Acid
CAS 144-90-1
Molecular Formula C4H9NO2
Quantity 250 mg
Synonym Propanoic acid, 3-amino-2-methyl-, 3-Amino-2-methylpropanoic acid, (±)-3-Amino-2-methylpropanoic acid, (±)-3-Amino-2-methylpropionic acid, (±)-3-Aminoisobutyric acid, (±)-β-Aminoisobutyric acid, 2-Methyl-3-aminopropionic acid, 2-Methyl-β-alanine, 3-Amino-2-methylpropionic acid, 3-Aminoisobutyric acid, DL-2-Methyl-β-alanine, DL-3-Amino-2-methylpropionic acid, DL-3-Aminoisobutyric acid, DL-3-amino-2-methylpropanoic acid, DL-β-Aminoisobutyric acid, α-Methyl-β-alanine, β-Aminoisobutyric acid
InChI Formula InChI=1S/C4H9NO2/c1-3(2-5)4(6)7/h3H,2,5H2,1H3,(H,6,7)
SMILES CC(CN)C(=O)O
IUPAC Name 3-amino-2-methylpropanoic acid
Molecular Weight (g/mol) 103.12
Formula Weight 103.0633
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.