missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-3-Aminoisobutyric Acid, TRC
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
Specifications
Specifications
| Chemical Name or Material | DL-3-Aminoisobutyric Acid |
| CAS | 144-90-1 |
| Molecular Formula | C4H9NO2 |
| Quantity | 2.5 g |
| Synonym | Propanoic acid, 3-amino-2-methyl-, 3-Amino-2-methylpropanoic acid, (±)-3-Amino-2-methylpropanoic acid, (±)-3-Amino-2-methylpropionic acid, (±)-3-Aminoisobutyric acid, (±)-β-Aminoisobutyric acid, 2-Methyl-3-aminopropionic acid, 2-Methyl-β-alanine, 3-Amino-2-methylpropionic acid, 3-Aminoisobutyric acid, DL-2-Methyl-β-alanine, DL-3-Amino-2-methylpropionic acid, DL-3-Aminoisobutyric acid, DL-3-amino-2-methylpropanoic acid, DL-β-Aminoisobutyric acid, α-Methyl-β-alanine, β-Aminoisobutyric acid |
| InChI Formula | InChI=1S/C4H9NO2/c1-3(2-5)4(6)7/h3H,2,5H2,1H3,(H,6,7) |
| SMILES | CC(CN)C(=O)O |
| IUPAC Name | 3-amino-2-methylpropanoic acid |
| Molecular Weight (g/mol) | 103.12 |
| Formula Weight | 103.0633 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction