missing translation for 'onlineSavingsMsg'
Learn More

DL-2-Phenylpropionaldehyde, 98%

Product Code. 10219503 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10219503 100 g Glass bottle 100g
10625952 25 g Glass bottle 25g
10229503 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10219503 Supplier Thermo Scientific Acros Supplier No. 130861000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 93-53-8 | C9H10O | 134.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93-53-8
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.18
MDL Number MFCD00006973
InChI Key IQVAERDLDAZARL-UHFFFAOYSA-N
Synonym 2-phenylpropionaldehyde, hydratropaldehyde, hydratropic aldehyde, hyacinthal, cumene aldehyde, hydratropa aldehyde, hydrotropic aldehyde, 2-phenyl-1-propanal, 2-phenyl propionaldehyde, aldehyd hydratropovy
PubChem CID 7146
IUPAC Name 2-phenylpropanal
SMILES CC(C=O)C1=CC=CC=C1

Specifications

Melting Point -60.0°C
Color Colorless
Density 1.0110g/mL
Boiling Point 202.0°C to 205.0°C
Flash Point 76°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH(C6H5)CHO
Refractive Index 1.5160 to 1.5240
Quantity 100 g
Beilstein 07, 305
Specific Gravity 1.011
Solubility Information Solubility in water: 3.4g/L (20°C).
Viscosity 2.16 mPa.s (20°C)
Formula Weight 134.18
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material DL-2-Phenylpropionaldehyde
Show More Show Less
compliance-icons
Product Identifier
  • DL-2-Phenylpropionaldehyde
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.