missing translation for 'onlineSavingsMsg'
Learn More

DL-2-Phenylglycine, 98%, Thermo Scientific™

Product Code. 11498960
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11498960 25 g 25g
11444617 100 g 100g
11454617 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11498960 Supplier Thermo Scientific Alfa Aesar Supplier No. B21129.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2835-06-5
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.17
MDL Number MFCD00064402
ChEBI CHEBI:55484

Specifications

Melting Point ∼290°C (sublimation)
Beilstein 3197862
Merck Index 14,7291
Synonym 2-phenylglycine, dl-2-phenylglycine, dl-alpha-phenylglycine, dl-phenylglycine, l-phenylglycine, amino phenyl acetic acid, amino-phenyl-acetic acid, alpha-phenylglycine, alpha-aminophenylacetic acid, aminophenylacetic acid
InChI Key ZGUNAGUHMKGQNY-UHFFFAOYNA-N
SMILES C1=CC=C(C=C1)C(C(=O)O)N
IUPAC Name 2-amino-2-phenylacetic acid
Quantity 25 g
PubChem CID 3866
Odor Odorless
Formula Weight 151.17
Percent Purity 98%
Chemical Name or Material DL-2-Phenylglycine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.