missing translation for 'onlineSavingsMsg'
Learn More

DL-2-Methylbutyryl chloride, 98%, Thermo Scientific Chemicals

Product Code. 10741252
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57526-28-0
Molecular Formula C5H9ClO
Molecular Weight (g/mol) 120.58
MDL Number MFCD00041726
InChI Key XRPVXVRWIDOORM-UHFFFAOYSA-N
Synonym 2-methylbutyryl chloride, dl-2-methylbutyryl chloride, butanoyl chloride, 2-methyl, ethylmethylacetylchloride, dl-2-methyl-butyryl chloride, rs-2-methylbutyryl chloride, 2-methyl-butyryl chloride, #, +-2-methyl butanoyl chloride
PubChem CID 93697
IUPAC Name 2-methylbutanoyl chloride
SMILES CCC(C)C(=O)Cl

Specifications

Color Colorless to Yellow
Density 0.9800g/mL
Boiling Point 116°C to 118°C
Flash Point 20°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula CH3CH2CH(CH3)COCl
Refractive Index 1.4160 to 1.418
Quantity 100 g
Specific Gravity 0.98
Formula Weight 120.58
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material DL-2-Methylbutyryl chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'