missing translation for 'onlineSavingsMsg'
Learn More

DL-2-Methylbutyryl chloride, 98%

Product Code. 10351111 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
1g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10351111 25 g Glass bottle 25g
10463385 1 g Glass bottle 1g
10095263 5 g Glass bottle 5g
10741252 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10351111 Supplier Thermo Scientific Acros Supplier No. 414150250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 57526-28-0 | C5H9ClO | 120.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57526-28-0
Molecular Formula C5H9ClO
Molecular Weight (g/mol) 120.58
MDL Number MFCD00041726
InChI Key XRPVXVRWIDOORM-UHFFFAOYSA-N
Synonym 2-methylbutyryl chloride, dl-2-methylbutyryl chloride, butanoyl chloride, 2-methyl, ethylmethylacetylchloride, dl-2-methyl-butyryl chloride, rs-2-methylbutyryl chloride, 2-methyl-butyryl chloride, #, +-2-methyl butanoyl chloride
PubChem CID 93697
IUPAC Name 2-methylbutanoyl chloride
SMILES CCC(C)C(=O)Cl

Specifications

Color Colorless to Yellow
Density 0.9800g/mL
Boiling Point 116°C to 118°C
Flash Point 20°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula CH3CH2CH(CH3)COCl
Refractive Index 1.4160 to 1.418
Quantity 25 g
Specific Gravity 0.98
Formula Weight 120.58
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material DL-2-Methylbutyryl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.